Order Entry
Export
Orders LinkContactUsLinkComponent
1-Tridecene 99%

1-Tridecene 99%

Health
Supplier:   Apollo Scientific
1-Tridecene 99%
Supplier:  Apollo Scientific
Certificates

5 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent.

  • CAS Number:2437-56-1
  • Formula:C₁₃H₂₆
  • MDL Number:MFCD00008976
  • Molecular Weight:182,35 g/mol
  • EINECS:219-443-8
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 66,60 €
    66,60 €
    1 * 5 g
       66,60 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 180,00 €
    180,00 €
    1 * 25 g
       180,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 701,00 €
    701,00 €
    1 * 100 g
       701,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 2 280,00 €
    2 280,00 €
    1 * 500 g
       2 280,00 €

    Specifications:
    New Product
    New Product