Order Entry
Export
Orders LinkContactUsLinkComponent
3-Bromopentane 99%

3-Bromopentane 99%

Flammable
Irritant
Supplier:   Apollo Scientific
3-Bromopentane 99%
Supplier:  Apollo Scientific
Certificates
3-Bromopentane 99%

5 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent.

  • CAS Number:1809-10-5
  • Formula:C₅H₁₁Br
  • MDL Number:MFCD00000158
  • Molecular Weight:151,04 g/mol
  • EINECS:217-314-0
  • UN:1993
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 23,20 €
    23,20 €
    1 * 1 g
       23,20 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 36,20 €
    36,20 €
    1 * 5 g
       36,20 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 63,70 €
    63,70 €
    1 * 25 g
       63,70 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 246,00 €
    246,00 €
    1 * 100 g
       246,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1 160,00 €
    1 160,00 €
    1 * 500 g
       1 160,00 €

    Specifications:
    New Product
    New Product