Order Entry
Export
Orders LinkContactUsLinkComponent
2-Methoxy-4-methylphenol 98%

2-Methoxy-4-methylphenol 98%

Supplier:   Apollo Scientific
2-Methoxy-4-methylphenol 98%
Supplier:  Apollo Scientific
Certificates
2-Methoxy-4-methylphenol 98%

4 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Creosol, 1-Hydroxy-2-methoxy-4-methylbenzene, 2-Methoxy-p-cresol, 2-Methoxy-4-methylphenol, 4-Hydroxy-3-methoxytoluene, 4-Methylguaiacol, 2-Hydroxy-5-methylanisol
  • CAS Number:93-51-6
  • Formula:C₈H₁₀O₂
  • MDL Number:MFCD00002378
  • Molecular Weight:138,16379 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 37,70 €
    37,70 €
    1 * 25 g
       37,70 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 60,80 €
    60,80 €
    1 * 100 g
       60,80 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 152,00 €
    152,00 €
    1 * 500 g
       152,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 300,00 €
    300,00 €
    1 * 1 kg
       300,00 €

    Specifications:
    New Product
    New Product