Order Entry
Export
Orders LinkContactUsLinkComponent
Dimethylphenylsilane 97%

Dimethylphenylsilane 97%

Flammable
Irritant
New Product
Supplier:   Apollo Scientific
Dimethylphenylsilane 97%
Supplier:  Apollo Scientific
New Product
Certificates
Dimethylphenylsilane 97%

5 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Dimethyl(phenyl)silane
  • CAS Number:766-77-8
  • Formula:C₈H₁₂Si
  • MDL Number:MFCD00008257
  • Molecular Weight:136,27 g/mol
  • EINECS:212-170-5
  • UN:1993
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,80 €
    21,80 €
    1 * 5 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 30,40 €
    30,40 €
    1 * 25 g
       30,40 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 116,00 €
    116,00 €
    1 * 100 g
       116,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 532,00 €
    532,00 €
    1 * 500 g
       532,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 2 760,00 €
    2 760,00 €
    1 * 2,5 kg
       2 760,00 €

    Specifications:
    New Product
    New Product