Undecan-2-Ol 98%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: Undecan-2-ol, (±)-Methyl nonyl carbinol, (±)-2-Hydroxy undecane, (±)-Nonyl methyl carbinol, 2-Undecanol
- Molecular Weight:172,31 g/mol
5 Options Available Below
Product Details & Documents
Synonyms: Undecan-2-ol, (±)-Methyl nonyl carbinol, (±)-2-Hydroxy undecane, (±)-Nonyl methyl carbinol, 2-Undecanol
- Molecular Weight:172,31 g/mol
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



