Order Entry
Export
Orders LinkContactUsLinkComponent
Dodecane 98%

Dodecane 98%

Bottle
Catalog # APOSOR95461-500G
Health
Supplier:   Apollo Scientific
Dodecane 98%
Catalog # APOSOR95461-500G
Supplier:  Apollo Scientific
CAS Number:  112-40-3
Dodecane 98%

Bottle

CAS Number:  112-40-3
Price
192,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Dodecane
  • CAS Number:112-40-3
  • Formula:C₁₂H₂₆
  • MDL Number:MFCD00008969
  • Molecular Weight:170,33 g/mol
  • EINECS:203-967-9
Specifications

Specifications

  • Pack type:
    Bottle

Specifications