Order Entry
Export
Orders LinkContactUsLinkComponent
Mandelic acid 99%

Mandelic acid 99%

New Product
Supplier:   Apollo Scientific
Mandelic acid 99%
Supplier:  Apollo Scientific
New Product
Certificates
Mandelic acid 99%

4 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  DL-Amygdalic Acid, Mandelic acid, DL-Mandelic acid, 2-Hydroxy-2-phenylacetic acid, (±)-2-Hydroxy-2-phenylacetic acid, DL-2-Hydroxy-2-phenylacetic acid
  • CAS Number:90-64-2
  • Formula:C₈H₈O₃
  • MDL Number:MFCD15143969
  • Molecular Weight:152,15 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 21,80 €
    21,80 €
    1 * 25 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 24,60 €
    24,60 €
    1 * 100 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 57,90 €
    57,90 €
    1 * 500 g
       57,90 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 254,00 €
    254,00 €
    1 * 2,5 kg
       254,00 €

    Specifications:
    New Product
    New Product