Order Entry
Export
Orders LinkContactUsLinkComponent
Ac-D-Pro-Oh 98%

Ac-D-Pro-Oh 98%

Supplier:   Apollo Scientific
Ac-D-Pro-Oh 98%
Supplier:  Apollo Scientific
Certificates

5 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  N-Acetyl-D-proline
  • CAS Number:59785-68-1
  • Formula:C₇H₁₁NO₃
  • MDL Number:MFCD20640695
  • Molecular Weight:157,17 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,80 €
    21,80 €
    1 * 5 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 23,20 €
    23,20 €
    1 * 25 g
       23,20 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 135,00 €
    135,00 €
    1 * 100 g
       135,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 429,00 €
    429,00 €
    1 * 500 g
       429,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 1 660,00 €
    1 660,00 €
    1 * 2,5 kg
       1 660,00 €

    Specifications:
    New Product
    New Product