Order Entry
Export
Orders LinkContactUsLinkComponent
2-Hexylthiophene 98%

2-Hexylthiophene 98%

Supplier:   Apollo Scientific
2-Hexylthiophene 98%
Supplier:  Apollo Scientific
Certificates
2-Hexylthiophene 98%

5 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent.

  • CAS Number:18794-77-9
  • Formula:C₁₀H₁₆S
  • MDL Number:MFCD00022535
  • Molecular Weight:168,3 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 24,60 €
    24,60 €
    1 * 5 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 59,40 €
    59,40 €
    1 * 25 g
       59,40 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 228,00 €
    228,00 €
    1 * 100 g
       228,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 804,00 €
    804,00 €
    1 * 500 g
       804,00 €

    Specifications:
    New Product
    New Product