Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Melamine
BP0000707697
Melamine
Print
Melamine
Bottle
Catalog # APOSBUP19758-5G
New Product
Supplier:
Apollo Scientific
Melamine
Catalog # APOSBUP19758-5G
Supplier:
Apollo Scientific
New Product
CAS Number:
Expand image
Return to Top
Melamine
Bottle
Specifications
Add to cart
Jump to:
Specifications
CAS Number:
Price
37,70 €
Log in
for availability
Unit of Measure
1 * 5 g
-
+
Add to cart
Return Policy
Product Details & Documents
Specifications
Specifications
Pack type:
Bottle
Specifications