Cinnamaldehyde ≥80%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Biochemical/life science applications.
Synonyms: trans-Cinnamaldehyde, 3-Phenylacrylaldehyde
- CAS Number:104-55-2
- Formula:C₉H₈O
- MDL Number:MFCD00007000
- Molecular Weight:132,16 g/mol
- EINECS:203-213-9
5 Options Available Below
Product Details & Documents
Usage: Biochemical/life science applications.
Synonyms: trans-Cinnamaldehyde, 3-Phenylacrylaldehyde
- CAS Number:104-55-2
- Formula:C₉H₈O
- MDL Number:MFCD00007000
- Molecular Weight:132,16 g/mol
- EINECS:203-213-9
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



