0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
3,6-Dihydroxyflavone
BP0000707383
3,6-Dihydroxyflavone
New Product
:
Apollo Scientific
3,6-Dihydroxyflavone
:
Apollo Scientific
New Product
3,6-Dihydroxyflavone
Product Family Options
Pack type
APOSBUP16420-100MG
Bottle
1 * 100 mg
1 * 100 mg - 407,00 €
407,00 €
-
+
1 * 100 mg
407,00 €
-
+
New Product
New Product
APOSBUP16420-500MG
Bottle
1 * 500 mg
1 * 500 mg - 909,00 €
909,00 €
-
+
1 * 500 mg
909,00 €
-
+
New Product
New Product