Tobramycin ≥98%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Biochemical/life science applications.
Synonyms: (2S,3R,4S,5S,6R)-4-Amino-2-(((1S,2S,3R,4S,6R)-4,6-diamino-3-(((2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxytetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,5-diol
- CAS Number:32986-56-4
- Formula:C₁₈H₃₇N₅O₉
- Molecular Weight:467,51 g/mol
4 Options Available Below
Product Details & Documents
Usage: Biochemical/life science applications.
Synonyms: (2S,3R,4S,5S,6R)-4-Amino-2-(((1S,2S,3R,4S,6R)-4,6-diamino-3-(((2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxytetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,5-diol
- CAS Number:32986-56-4
- Formula:C₁₈H₃₇N₅O₉
- Molecular Weight:467,51 g/mol
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



