Order Entry
Export
Orders LinkContactUsLinkComponent
Estradiol ≥98%

Estradiol ≥98%

New Product
Supplier:   Apollo Scientific
Estradiol ≥98%
Supplier:  Apollo Scientific
New Product
Certificates
Estradiol ≥98%

2 Options Available Below

Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  beta-Estradiol, 17β-Estradiol
  • CAS Number:50-28-2
  • Formula:C₁₈H₂₄O₂
  • Molecular Weight:272,38 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 300,00 €
    300,00 €
    1 * 5 g
       300,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 442,00 €
    442,00 €
    1 * 10 g
       442,00 €

    Specifications:
    New Product
    New Product