Order Entry
Export
Orders LinkContactUsLinkComponent
Phenolphthalein indicator

Phenolphthalein indicator

New Product
Supplier:   Apollo Scientific
Phenolphthalein indicator
Supplier:  Apollo Scientific
New Product
Certificates
Phenolphthalein indicator

4 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
  • CAS Number:77-09-8
  • Formula:C₂₀H₁₄O₄
  • MDL Number:MFCD00005913
  • Molecular Weight:318,32 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 21,80 €
    21,80 €
    1 * 25 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 24,60 €
    24,60 €
    1 * 100 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 57,90 €
    57,90 €
    1 * 500 g
       57,90 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 254,00 €
    254,00 €
    1 * 2,5 kg
       254,00 €

    Specifications:
    New Product
    New Product