Ac-Tyr(Tbu)-OH 98%
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Organic building block/reagent.
Synonyms: Ac-Tyr(tBu)-OH
- CAS Number:201292-99-1
- Formula:C₁₅H₂₁NO₄
- MDL Number:MFCD24532698
- Molecular Weight:279,33 g/mol
4 Options Available Below
Product Details & Documents
Usage: Organic building block/reagent.
Synonyms: Ac-Tyr(tBu)-OH
- CAS Number:201292-99-1
- Formula:C₁₅H₂₁NO₄
- MDL Number:MFCD24532698
- Molecular Weight:279,33 g/mol
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



