Order Entry
Export
Orders LinkContactUsLinkComponent
 

Bronopol ≥98%

Bottle
  APOSBUP19183-1ML
New Product
Bronopol ≥98%
  APOSBUP19183-1ML
New Product
 :  52-51-7
Bronopol ≥98%

Bottle

 :  52-51-7
 
63,70 €
   
1 * 1 mL
 

Usage: Biochemical/life science applications.

 :  2-Bromo-2-nitro-1,3-propanediol, 2-Bromo-2-nitropropane-1,3-diol
  •  :52-51-7
  •  :C₃H₆BrNO₄
  •  :199,99 g/mol
 

 

  • Pack type:
    Bottle