Folic acid ≥95%
Bottle
Catalog # APOSBUP18249-500MG
New Product
Supplier: Apollo Scientific
Folic acid ≥95%
Catalog # APOSBUP18249-500MG
Supplier: Apollo Scientific
New Product
CAS Number: 59-30-3
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications.
Synonyms: Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9
- CAS Number:59-30-3
- Formula:C₁₉H₁₉N₇O₆
- Molecular Weight:441,4 g/mol
Specifications
Specifications
- Pack type:Bottle



