Order Entry
Export
Orders LinkContactUsLinkComponent
L-Thyroxine ≥95%

L-Thyroxine ≥95%

New Product
Supplier:   Apollo Scientific
L-Thyroxine ≥95%
Supplier:  Apollo Scientific
New Product
Certificates
L-Thyroxine ≥95%

2 Options Available Below

Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  (S)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid
  • CAS Number:51-48-9
  • Formula:C₁₅H₁₁I₄NO₄
  • Molecular Weight:776,87 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 40,60 €
    40,60 €
    1 * 200 mg
       40,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 68,10 €
    68,10 €
    1 * 500 mg
       68,10 €

    Specifications:
    New Product
    New Product