Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
N-Cycloheptyl-2-(4,5-dihydro-1H-imidazol-2-ylsulphanyl)acetamide hydrochloride
BP0000704612
ICCB-19 hydrochloride
Print
ICCB-19 hydrochloride
New Product
Supplier:
Apollo Scientific
ICCB-19 hydrochloride
Supplier:
Apollo Scientific
New Product
Certificates
Expand image
Return to Top
ICCB-19 hydrochloride
Product Family Options
Add to cart
Jump to:
Product Family Options
About this item
3 Options Available Below
Shop Now
Return Policy
Product Details & Documents
Product Family Options
Product Information
Pack type
Availability
Price
APOSBUP15604-50MG
Bottle
Log in
for availability
Log in
for availability
1 * 50 mg
1 * 50 mg - 189,00 €
189,00 €
-
+
1 * 50 mg
189,00 €
-
+
Specifications:
New Product
New Product
APOSBUP15604-100MG
Bottle
Log in
for availability
Log in
for availability
1 * 100 mg
1 * 100 mg - 283,00 €
283,00 €
-
+
1 * 100 mg
283,00 €
-
+
Specifications:
New Product
New Product
APOSBUP15604-200MG
Bottle
Log in
for availability
Log in
for availability
1 * 200 mg
1 * 200 mg - 419,00 €
419,00 €
-
+
1 * 200 mg
419,00 €
-
+
Specifications:
New Product
New Product