(±)-10-Hydroxycamptothecin ≥98%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Biochemical/life science applications.
Synonyms: 1-HCPT, 10-Hydroxycamptothecin, 4-Ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, NSC107124
- CAS Number:64439-81-2
- Formula:C₂₀H₁₆N₂O₅
- Molecular Weight:364,35 g/mol
4 Options Available Below
Product Details & Documents
Usage: Biochemical/life science applications.
Synonyms: 1-HCPT, 10-Hydroxycamptothecin, 4-Ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, NSC107124
- CAS Number:64439-81-2
- Formula:C₂₀H₁₆N₂O₅
- Molecular Weight:364,35 g/mol
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



