0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
3-(Benzylthio)-5-(phenoxymethyl)-4-phenyl-4H-1,2,4-triazole
BP0000704207
MIND4-19
New Product
:
Apollo Scientific
MIND4-19
:
Apollo Scientific
New Product
MIND4-19
Product Family Options
Pack type
APOSBUP10873-25MG
Bottle
1 * 25 mg
1 * 25 mg - 176,00 €
176,00 €
-
+
1 * 25 mg
176,00 €
-
+
New Product
New Product
APOSBUP10873-50MG
Bottle
1 * 50 mg
1 * 50 mg - 284,00 €
284,00 €
-
+
1 * 50 mg
284,00 €
-
+
New Product
New Product
APOSBUP10873-100MG
Bottle
1 * 100 mg
1 * 100 mg - 456,00 €
456,00 €
-
+
1 * 100 mg
456,00 €
-
+
New Product
New Product
APOSBUP10873-200MG
Bottle
1 * 200 mg
1 * 200 mg - 643,00 €
643,00 €
-
+
1 * 200 mg
643,00 €
-
+
New Product
New Product