0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Chromone
BP0000703969
Chromone
New Product
:
Apollo Scientific
Chromone
:
Apollo Scientific
New Product
Chromone
Product Family Options
Pack type
APOSOR22414-1G
Bottle
1 * 1 g
1 * 1 g - 21,80 €
21,80 €
-
+
1 * 1 g
21,80 €
-
+
New Product
New Product
APOSOR22414-5G
Bottle
1 * 5 g
1 * 5 g - 24,60 €
24,60 €
-
+
1 * 5 g
24,60 €
-
+
New Product
New Product
APOSOR22414-25G
Bottle
1 * 25 g
1 * 25 g - 94,10 €
94,10 €
-
+
1 * 25 g
94,10 €
-
+
New Product
New Product
APOSOR22414-100G
Bottle
1 * 100 g
1 * 100 g - 372,00 €
372,00 €
-
+
1 * 100 g
372,00 €
-
+
New Product
New Product
APOSOR22414-500G
Bottle
1 * 500 g
1 * 500 g - 1 560,00 €
1 560,00 €
-
+
1 * 500 g
1 560,00 €
-
+
New Product
New Product