Order Entry
Export
Orders LinkContactUsLinkComponent
 

7-Hydroxycoumarin 98%

Irritant
New Product
Supplier:   Apollo Scientific
7-Hydroxycoumarin 98%
Supplier:  Apollo Scientific
New Product
Certificates
7-Hydroxycoumarin 98%

3 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone
  • CAS Number:93-35-6
  • Formula:C₉H₆O₃
  • MDL Number:MFCD00006878
  • Molecular Weight:162,14213 g/mol
  • EINECS:202-240-3
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 42,00 €
    42,00 €
    1 * 25 g
       42,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 106,00 €
    106,00 €
    1 * 100 g
       106,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 519,00 €
    519,00 €
    1 * 500 g
       519,00 €

    Specifications:
    New Product
    New Product