Order Entry
Export
Orders LinkContactUsLinkComponent
 

10-Nonadecanamine 98%

New Product
Supplier:   Apollo Scientific
10-Nonadecanamine 98%
Supplier:  Apollo Scientific
New Product
Certificates
10-Nonadecanamine 98%

4 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent.

  • CAS Number:3241-23-4
  • Formula:C₁₉H₄₁N
  • Molecular Weight:283,54 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 56,50 €
    56,50 €
    1 * 250 mg
       56,50 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 152,00 €
    152,00 €
    1 * 1 g
       152,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 533,00 €
    533,00 €
    1 * 5 g
       533,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 2 560,00 €
    2 560,00 €
    1 * 25 g
       2 560,00 €

    Specifications:
    New Product
    New Product