0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
3,6-Dihydroxyflavone
BP0000700986
3,6-Dihydroxyflavone
New Product
:
Apollo Scientific
3,6-Dihydroxyflavone
:
Apollo Scientific
New Product
3,6-Dihydroxyflavone
Product Family Options
Pack type
APOSOR1027705-100M
Bottle
1 * 100 mg
1 * 100 mg - 31,90 €
31,90 €
-
+
1 * 100 mg
31,90 €
-
+
New Product
New Product
APOSOR1027705-250M
Bottle
1 * 250 mg
1 * 250 mg - 55,00 €
55,00 €
-
+
1 * 250 mg
55,00 €
-
+
New Product
New Product
APOSOR1027705-1G
Bottle
1 * 1 g
1 * 1 g - 148,00 €
148,00 €
-
+
1 * 1 g
148,00 €
-
+
New Product
New Product
APOSOR1027705-5G
Bottle
1 * 5 g
1 * 5 g - 516,00 €
516,00 €
-
+
1 * 5 g
516,00 €
-
+
New Product
New Product
APOSOR1027705-25G
Bottle
1 * 25 g
1 * 25 g - 2 370,00 €
2 370,00 €
-
+
1 * 25 g
2 370,00 €
-
+
New Product
New Product