Order Entry
Export
Orders LinkContactUsLinkComponent
Cyclohexanone 2,4-Dinitrophenylhydrazone 99%

Cyclohexanone 2,4-Dinitrophenylhydrazone 99%

New Product
Supplier:   Apollo Scientific
Cyclohexanone 2,4-Dinitrophenylhydrazone 99%
Supplier:  Apollo Scientific
New Product
Certificates
Cyclohexanone 2,4-Dinitrophenylhydrazone 99%

5 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent

  • CAS Number:1589-62-4
  • Formula:C₁₂H₁₄N₄O₄
  • Molecular Weight:278,26 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 26,10 €
    26,10 €
    1 * 100 mg
       26,10 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 55,00 €
    55,00 €
    1 * 250 mg
       55,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 138,00 €
    138,00 €
    1 * 1 g
       138,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 659,00 €
    659,00 €
    1 * 5 g
       659,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 3 230,00 €
    3 230,00 €
    1 * 25 g
       3 230,00 €

    Specifications:
    New Product
    New Product