AC-55541 98%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Organic building block/reagent
Synonyms: (E)-N-(2-(2-(1-(3-Bromophenyl)ethylidene)hydrazineyl)-2-oxo-1-(4-oxo-3,4-dihydrophthalazin-1-yl)ethyl)benzamide
- CAS Number:916170-19-9
- Formula:C₂₅H₂₀BrN₅O₃
- Molecular Weight:518,36 g/mol
4 Options Available Below
Product Details & Documents
Usage: Organic building block/reagent
Synonyms: (E)-N-(2-(2-(1-(3-Bromophenyl)ethylidene)hydrazineyl)-2-oxo-1-(4-oxo-3,4-dihydrophthalazin-1-yl)ethyl)benzamide
- CAS Number:916170-19-9
- Formula:C₂₅H₂₀BrN₅O₃
- Molecular Weight:518,36 g/mol
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



