Order Entry
Export
Orders LinkContactUsLinkComponent
 

Silica 99, 8%, fumed

Irritant
New Product
Supplier:   Apollo Scientific
Silica 99, 8%, fumed
Supplier:  Apollo Scientific
New Product
Certificates
Silica 99, 8%, fumed

3 Options Available Below

Product Details & Documents

Hydrophobic type R974, Specific surface area 170 ±20 m²/g; Particle size: 12 nm.

  • Usage: Organic building block/reagent
  • CAS Number:112945-52-5
  • Formula:SiO₂
  • Molecular Weight:60,08 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 21,80 €
    21,80 €
    1 * 100 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 39,10 €
    39,10 €
    1 * 500 g
       39,10 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 187,00 €
    187,00 €
    1 * 2,5 kg
       187,00 €

    Specifications:
    New Product
    New Product