Order Entry
Export
Orders LinkContactUsLinkComponent
1-Allylimidazole 98%

1-Allylimidazole 98%

Supplier:   Apollo Scientific
1-Allylimidazole 98%
Supplier:  Apollo Scientific
Certificates
1-Allylimidazole 98%

5 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  1-Allyl-1H-imidazole
  • CAS Number:31410-01-2
  • Formula:C₆H₈N₂
  • MDL Number:MFCD00037894
  • Molecular Weight:108,14 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,80 €
    21,80 €
    1 * 5 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 46,40 €
    46,40 €
    1 * 25 g
       46,40 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 154,00 €
    154,00 €
    1 * 100 g
       154,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 657,00 €
    657,00 €
    1 * 500 g
       657,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 3 160,00 €
    3 160,00 €
    1 * 2,5 kg
       3 160,00 €

    Specifications:
    New Product
    New Product