Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
2-Mercaptobenzothiazole
BP0000699445
Print
2-Mercaptobenzothiazole
New Product
Supplier:
Apollo Scientific
2-Mercaptobenzothiazole
Supplier:
Apollo Scientific
New Product
Certificates
Return to Top
2-Mercaptobenzothiazole
Product Family Options
Jump to:
Product Family Options
About this item
3 Options Available Below
Shop Now
Return Policy
Product Details & Documents
Product Family Options
Product Information
Pack type
Availability
Price
APOSBUP19642-1G
Bottle
Log in
for availability
Log in
for availability
1 * 1 g
1 * 1 g - 40,60 €
40,60 €
-
+
1 * 1 g
40,60 €
-
+
Specifications:
New Product
New Product
APOSBUP19642-5G
Bottle
Log in
for availability
Log in
for availability
1 * 5 g
1 * 5 g - 89,80 €
89,80 €
-
+
1 * 5 g
89,80 €
-
+
Specifications:
New Product
New Product
APOSBUP19642-10G
Bottle
Log in
for availability
Log in
for availability
1 * 10 g
1 * 10 g - 126,00 €
126,00 €
-
+
1 * 10 g
126,00 €
-
+
Specifications:
New Product
New Product