Order Entry
Export
Orders LinkContactUsLinkComponent
 

Dexamethasone acetate ≥98%

New Product
Supplier:   Apollo Scientific
Dexamethasone acetate ≥98%
Supplier:  Apollo Scientific
New Product
Certificates
Dexamethasone acetate ≥98%

3 Options Available Below

Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  11β,17α-Dihydroxy-21-(ethanoyloxy)-9-fluor-16α-methyl-1,4-pregnadien-3,20-dion, Dexamethasone acetate
  • CAS Number:1177-87-3
  • Formula:C₂₄H₃₁FO₆
  • Molecular Weight:434,5 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 57,90 €
    57,90 €
    1 * 500 mg
       57,90 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 98,40 €
    98,40 €
    1 * 1 g
       98,40 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 313,00 €
    313,00 €
    1 * 10 g
       313,00 €

    Specifications:
    New Product
    New Product