Order Entry
Export
Orders LinkContactUsLinkComponent
Thymidine ≥95%

Thymidine ≥95%

Bottle
Catalog # APOSBUP18243-500MG
New Product
Supplier:   Apollo Scientific
Thymidine ≥95%
Catalog # APOSBUP18243-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  50-89-5
Thymidine ≥95%

Bottle

CAS Number:  50-89-5
Price
57,90 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine
  • CAS Number:50-89-5
  • Formula:C₁₀H₁₄N₂O₅
  • Molecular Weight:242,23 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications