Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
7-Hydroxycoumarin
BP0000698976
Umbelliferone
Print
Umbelliferone
New Product
Supplier:
Apollo Scientific
Umbelliferone
Supplier:
Apollo Scientific
New Product
Certificates
Expand image
Return to Top
Umbelliferone
Product Family Options
Add to cart
Jump to:
Product Family Options
About this item
3 Options Available Below
Shop Now
Return Policy
Product Details & Documents
Product Family Options
Product Information
Pack type
Availability
Price
APOSBUP14594-50MG
Bottle
Log in
for availability
Log in
for availability
1 * 50 mg
1 * 50 mg - 39,10 €
39,10 €
-
+
1 * 50 mg
39,10 €
-
+
Specifications:
New Product
New Product
APOSBUP14594-5G
Bottle
Log in
for availability
Log in
for availability
1 * 5 g
1 * 5 g - 348,00 €
348,00 €
-
+
1 * 5 g
348,00 €
-
+
Specifications:
New Product
New Product
APOSBUP14594-10G
Bottle
Log in
for availability
Log in
for availability
1 * 10 g
1 * 10 g - 529,00 €
529,00 €
-
+
1 * 10 g
529,00 €
-
+
Specifications:
New Product
New Product