0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
(S)-10-hydroxycamptothecin
BP0000698916
(S)-10-Hydroxycamptothecin
New Product
:
Apollo Scientific
(S)-10-Hydroxycamptothecin
:
Apollo Scientific
New Product
(S)-10-Hydroxycamptothecin
Product Family Options
Pack type
APOSBUP13827-25MG
Bottle
1 * 25 mg
1 * 25 mg - 56,50 €
56,50 €
-
+
1 * 25 mg
56,50 €
-
+
New Product
New Product
APOSBUP13827-50MG
Bottle
1 * 50 mg
1 * 50 mg - 76,70 €
76,70 €
-
+
1 * 50 mg
76,70 €
-
+
New Product
New Product
APOSBUP13827-100MG
Bottle
1 * 100 mg
1 * 100 mg - 108,00 €
108,00 €
-
+
1 * 100 mg
108,00 €
-
+
New Product
New Product
APOSBUP13827-500MG
Bottle
1 * 500 mg
1 * 500 mg - 251,00 €
251,00 €
-
+
1 * 500 mg
251,00 €
-
+
New Product
New Product
APOSBUP13827-1G
Bottle
1 * 1 g
1 * 1 g - 338,00 €
338,00 €
-
+
1 * 1 g
338,00 €
-
+
New Product
New Product