Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
2,2'-((((2-Fluoro-1,3-phenylene)bis(methylene))bis(oxy))bis(4,1-phenylene))bis(N-isopropyl-1H-benzo[d]imidazole-6-carboximidamide)
BP0000698028
DB2313
Print
DB2313
New Product
Supplier:
Apollo Scientific
DB2313
Supplier:
Apollo Scientific
New Product
Certificates
Expand image
Return to Top
DB2313
Product Family Options
Add to cart
Jump to:
Product Family Options
About this item
2 Options Available Below
Shop Now
Return Policy
Product Details & Documents
Product Family Options
Product Information
Pack type
Availability
Price
APOSBUP00943-25MG
Bottle
Log in
for availability
Log in
for availability
1 * 25 mg
1 * 25 mg - 579,00 €
579,00 €
-
+
1 * 25 mg
579,00 €
-
+
Specifications:
New Product
New Product
APOSBUP00943-50MG
Bottle
Log in
for availability
Log in
for availability
1 * 50 mg
1 * 50 mg - 856,00 €
856,00 €
-
+
1 * 50 mg
856,00 €
-
+
Specifications:
New Product
New Product