Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
tri-Calcium phosphate
BP0000696773
Calcium phosphate
Print
Calcium phosphate
New Product
Supplier:
Apollo Scientific
Calcium phosphate
Supplier:
Apollo Scientific
New Product
Certificates
Expand image
Return to Top
Calcium phosphate
Product Family Options
Add to cart
Jump to:
Product Family Options
About this item
3 Options Available Below
Shop Now
Return Policy
Product Details & Documents
Product Family Options
Product Information
Pack type
Availability
Price
APOSIN1392-5G
Bottle
Log in
for availability
Log in
for availability
1 * 5 g
1 * 5 g - 21,80 €
21,80 €
-
+
1 * 5 g
21,80 €
-
+
Specifications:
New Product
New Product
APOSIN1392-25G
Bottle
Log in
for availability
Log in
for availability
1 * 25 g
1 * 25 g - 36,20 €
36,20 €
-
+
1 * 25 g
36,20 €
-
+
Specifications:
New Product
New Product
APOSIN1392-100G
Bottle
Log in
for availability
Log in
for availability
1 * 100 g
1 * 100 g - 86,90 €
86,90 €
-
+
1 * 100 g
86,90 €
-
+
Specifications:
New Product
New Product