Order Entry
Export
Orders LinkContactUsLinkComponent
DL-Alanine ≥98%

DL-Alanine ≥98%

Bottle
Catalog # APOSBUP19841-5G
New Product
Supplier:   Apollo Scientific
DL-Alanine ≥98%
Catalog # APOSBUP19841-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  302-72-7
DL-Alanine ≥98%

Bottle

CAS Number:  302-72-7
Price
37,70 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  (±)-Alanine, DL-α-Alanine, H-DL-Ala-OH, α-Alanine
  • CAS Number:302-72-7
  • Formula:C₃H₇NO₂
  • Molecular Weight:89,09 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications