Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Propanediol ≥98%

1,3-Propanediol ≥98%

New Product
Supplier:   Apollo Scientific
1,3-Propanediol ≥98%
Supplier:  Apollo Scientific
New Product
Certificates
1,3-Propanediol ≥98%

2 Options Available Below

Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  1,3-Propylene glycol, 1,3-Dihydroxy propane
  • CAS Number:504-63-2
  • Formula:C₃H₈O₂
  • Molecular Weight:76,09 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 60,80 €
    60,80 €
    1 * 5 g
       60,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 86,90 €
    86,90 €
    1 * 10 g
       86,90 €

    Specifications:
    New Product
    New Product