Order Entry
Export
Orders LinkContactUsLinkComponent
Malic acid ≥95%

Malic acid ≥95%

Bottle
Catalog # APOSBUP18982-1ML
New Product
Supplier:   Apollo Scientific
Malic acid ≥95%
Catalog # APOSBUP18982-1ML
Supplier:  Apollo Scientific
New Product
CAS Number:  6915-15-7
Malic acid ≥95%

Bottle

CAS Number:  6915-15-7
Price
37,70 €
Log in for availability
1 * 1 mL
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  DL-Hydroxysuccinic acid, (±)-Malic acid, (±)-Hydroxy-1,4-butanedioic acid, (±)-Hydroxysuccinic acid, Hydroxybutanedioic acid, 2-Hydroxysuccinic acid
  • CAS Number:6915-15-7
  • Formula:C₄H₆O₅
  • Molecular Weight:134,09 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications