Order Entry
Export
Orders LinkContactUsLinkComponent
Adenosine ≥98%

Adenosine ≥98%

New Product
Supplier:   Apollo Scientific
Adenosine ≥98%
Supplier:  Apollo Scientific
New Product
Certificates
Adenosine ≥98%

3 Options Available Below

Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
  • CAS Number:58-61-7
  • Formula:C₁₀H₁₃N₅O₄
  • Molecular Weight:267,24 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 59,40 €
    59,40 €
    1 * 500 mg
       59,40 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 71,00 €
    71,00 €
    1 * 1 g
       71,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 112,00 €
    112,00 €
    1 * 5 g
       112,00 €

    Specifications:
    New Product
    New Product