Phenacetin ≥98%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Biochemical/life science applications
Synonyms: Acetphenetidin, Acetophenetidin, Phenacetin, N-(4-Ethoxyphenyl)acetamide, 4-Acetophenetidide
- CAS Number:62-44-2
- Formula:C₁₀H₁₃NO₂
- Molecular Weight:179,22 g/mol
- EINECS:200-533-0
2 Options Available Below
Product Details & Documents
Usage: Biochemical/life science applications
Synonyms: Acetphenetidin, Acetophenetidin, Phenacetin, N-(4-Ethoxyphenyl)acetamide, 4-Acetophenetidide
- CAS Number:62-44-2
- Formula:C₁₀H₁₃NO₂
- Molecular Weight:179,22 g/mol
- EINECS:200-533-0



