Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
L-Methionine
BP0000696533
L-Methionine
Print
L-Methionine
New Product
Supplier:
Apollo Scientific
L-Methionine
Supplier:
Apollo Scientific
New Product
Certificates
Expand image
Return to Top
L-Methionine
Product Family Options
Add to cart
Jump to:
Product Family Options
About this item
2 Options Available Below
Shop Now
Return Policy
Product Details & Documents
Product Family Options
Product Information
Pack type
Availability
Price
APOSBUP18381-500MG
Bottle
Log in
for availability
Log in
for availability
1 * 500 mg
1 * 500 mg - 52,10 €
52,10 €
-
+
1 * 500 mg
52,10 €
-
+
Specifications:
New Product
New Product
APOSBUP18381-1G
Bottle
Log in
for availability
Log in
for availability
1 * 1 g
1 * 1 g - 62,30 €
62,30 €
-
+
1 * 1 g
62,30 €
-
+
Specifications:
New Product
New Product