Order Entry
Export
Orders LinkContactUsLinkComponent
O4I1 ≥95%

O4I1 ≥95%

New Product
Supplier:   Apollo Scientific
O4I1 ≥95%
Supplier:  Apollo Scientific
New Product
Certificates

4 Options Available Below

Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  4-[(4-Methoxyphenyl)methoxy]benzeneacetonitrile, 2-(4-((4-Methoxybenzyl)oxy)phenyl)acetonitrile, O4I1
  • CAS Number:175135-47-4
  • Formula:C₁₆H₁₅NO₂
  • Molecular Weight:253,3 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 66,60 €
    66,60 €
    1 * 25 mg
       66,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 102,00 €
    102,00 €
    1 * 50 mg
       102,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 176,00 €
    176,00 €
    1 * 100 mg
       176,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 303,00 €
    303,00 €
    1 * 200 mg
       303,00 €

    Specifications:
    New Product
    New Product