Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
1,1'-(((Ethane-1,2-diylbis(oxy))bis(4,1-phenylene))bis(methylene))bis(4-(dimethylamino)pyridin-1-ium) bromide
BP0000695416
Print
EB-3D
New Product
Supplier:
Apollo Scientific
EB-3D
Supplier:
Apollo Scientific
New Product
Certificates
Return to Top
EB-3D
Product Family Options
Jump to:
Product Family Options
About this item
3 Options Available Below
Shop Now
Return Policy
Product Details & Documents
Product Family Options
Product Information
Pack type
Availability
Price
APOSBUP03990-25MG
Bottle
Log in
for availability
Log in
for availability
1 * 25 mg
1 * 25 mg - 631,00 €
631,00 €
-
+
1 * 25 mg
631,00 €
-
+
Specifications:
New Product
New Product
APOSBUP03990-50MG
Bottle
Log in
for availability
Log in
for availability
1 * 50 mg
1 * 50 mg - 876,00 €
876,00 €
-
+
1 * 50 mg
876,00 €
-
+
Specifications:
New Product
New Product
APOSBUP03990-100MG
Bottle
Log in
for availability
Log in
for availability
1 * 100 mg
1 * 100 mg - 1 240,00 €
1 240,00 €
-
+
1 * 100 mg
1 240,00 €
-
+
Specifications:
New Product
New Product