Order Entry
Export
Orders LinkContactUsLinkComponent
8-Bromo-1,7-naphthyridine 96%

8-Bromo-1,7-naphthyridine 96%

New Product
Supplier:   Apollo Scientific
8-Bromo-1,7-naphthyridine 96%
Supplier:  Apollo Scientific
New Product
Certificates
8-Bromo-1,7-naphthyridine 96%

4 Options Available Below

Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:63845-72-7
  • Formula:C₈H₅BrN₂
  • MDL Number:MFCD12401002
  • Molecular Weight:209,046 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 189,00 €
    189,00 €
    1 * 250 mg
       189,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 377,00 €
    377,00 €
    1 * 1 g
       377,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 1 830,00 €
    1 830,00 €
    1 * 5 g
       1 830,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 3 160,00 €
    3 160,00 €
    1 * 10 g
       3 160,00 €

    Specifications:
    New Product
    New Product