Order Entry
Export
Orders LinkContactUsLinkComponent
Palmitic acid 98%

Palmitic acid 98%

Irritant
New Product
Supplier:   Apollo Scientific
Palmitic acid 98%
Supplier:  Apollo Scientific
New Product
Certificates
Palmitic acid 98%

4 Options Available Below

Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  Hexadecanoic acid, Hexadecylic acid
  • CAS Number:57-10-3
  • Formula:C₁₆H₃₂O₂
  • MDL Number:MFCD00002747
  • Molecular Weight:256,4268 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 21,80 €
    21,80 €
    1 * 25 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 24,60 €
    24,60 €
    1 * 100 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 57,90 €
    57,90 €
    1 * 500 g
       57,90 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 254,00 €
    254,00 €
    1 * 2,5 kg
       254,00 €

    Specifications:
    New Product
    New Product