Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
TCS OX2 29
BP0000691836
TCS-OX2-29
Print
TCS-OX2-29
New Product
Supplier:
Apollo Scientific
TCS-OX2-29
Supplier:
Apollo Scientific
New Product
Certificates
Expand image
Return to Top
TCS-OX2-29
Product Family Options
Add to cart
Jump to:
Product Family Options
About this item
3 Options Available Below
Shop Now
Return Policy
Product Details & Documents
Product Family Options
Product Information
Pack type
Availability
Price
APOSBUP05857-25MG
Bottle
Log in
for availability
Log in
for availability
1 * 25 mg
1 * 25 mg - 241,00 €
241,00 €
-
+
1 * 25 mg
241,00 €
-
+
Specifications:
New Product
New Product
APOSBUP05857-50MG
Bottle
Log in
for availability
Log in
for availability
1 * 50 mg
1 * 50 mg - 355,00 €
355,00 €
-
+
1 * 50 mg
355,00 €
-
+
Specifications:
New Product
New Product
APOSBUP05857-100MG
Bottle
Log in
for availability
Log in
for availability
1 * 100 mg
1 * 100 mg - 633,00 €
633,00 €
-
+
1 * 100 mg
633,00 €
-
+
Specifications:
New Product
New Product