Order Entry
Export
Orders LinkContactUsLinkComponent
 

Fast red ITR base 98%

New Product
Supplier:   Apollo Scientific
Fast red ITR base 98%
Supplier:  Apollo Scientific
New Product
Certificates
Fast red ITR base 98%

5 Options Available Below

Product Details & Documents

Usage: Organic building block/ reagent.

  • Non-hazardous
Synonyms:  Azoic Diazo Component 42, N1,N1-Diethyl-4-methoxymetanilamide, Fast Red ITR, 3-Amino-N,N-diethyl-4-methoxybenzenesulphonamide
  • CAS Number:97-35-8
  • Formula:C₁₁H₁₈N₂O₃S
  • Molecular Weight:258,34 g/mol
  • EINECS:202-575-5
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,80 €
    21,80 €
    1 * 5 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 24,60 €
    24,60 €
    1 * 25 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 34,80 €
    34,80 €
    1 * 100 g
       34,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 165,00 €
    165,00 €
    1 * 500 g
       165,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 804,00 €
    804,00 €
    1 * 2,5 kg
       804,00 €

    Specifications:
    New Product
    New Product