Order Entry
Export
Orders LinkContactUsLinkComponent
2,2-Dimethoxybiphenyl 97%

2,2-Dimethoxybiphenyl 97%

New Product
Supplier:   Apollo Scientific
2,2-Dimethoxybiphenyl 97%
Supplier:  Apollo Scientific
New Product
Certificates
2,2-Dimethoxybiphenyl 97%

5 Options Available Below

Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:4877-93-4
  • Formula:C₁₄H₁₄O₂
  • Molecular Weight:214,26 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,80 €
    21,80 €
    1 * 5 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 63,70 €
    63,70 €
    1 * 25 g
       63,70 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 194,00 €
    194,00 €
    1 * 100 g
       194,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 731,00 €
    731,00 €
    1 * 500 g
       731,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 3 300,00 €
    3 300,00 €
    1 * 2,5 kg
       3 300,00 €

    Specifications:
    New Product
    New Product